siRNA Details
| Gene Symbol: Smad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008540.2 | NP_032566.2 | 3, 4 | 865 | ||
| GenBank mRNA | AB221585.1 | - | 2, 3 | 425 | ||
| AK004804.1 | - | 2, 3 | 229 | |||
| AY493561.1 | - | 2, 3 | 425 | |||
| BC046584.1 | - | 3, 4 | 865 | 6313280 | ||
| GU072917.1 | - | 2, 3 | 425 | |||
| GU072918.1 | - | 2, 3 | 425 | |||
| GU072919.1 | - | 2, 3 | 425 | |||
| GU072920.1 | - | 2, 3 | 425 | |||
| U79748.1 | - | 2, 3 | 745 | |||

Smad4
Genomic Map