siRNA Details
| Gene Symbol: Il17rd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134437.3 | NP_602319.1 | 3 | 381 | ||
| GenBank mRNA | AF459444.1 | - | 3 | 300 | ||
| AF494210.1 | - | 3 | 441 | |||
| AK133233.1 | - | 5 | 544 | |||
| BC066804.1 | - | 3 | 303 | |||
| BC138629.1 | - | 3 | 336 | 8861649 | ||

Il17rd
Genomic Map