siRNA Details
| Gene Symbol: Man1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010763.2 | NP_034893.1 | 4 | 1294 | ||
| GenBank mRNA | AK153336.1 | - | 4 | 1294 | ||
| BC049121.1 | - | 4 | 698 | 6813258 | ||
| BC068192.1 | - | 4 | 698 | 6813258 | ||
| U03457.1 | - | 4 | 1272 | |||
| U03458.1 | - | 4 | 1272 | |||

Man1a2
Genomic Map