siRNA Details
| Gene Symbol: Mical1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164433.1 | NP_001157905.1 | 12 | 1813 | ||
| NM_138315.2 | NP_612188.1 | 13 | 2032 | |||
| GenBank mRNA | AK154665.1 | - | 13 | 1972 | ||
| AK154930.1 | - | 13 | 2012 | |||
| AK157196.1 | - | 13 | 1969 | |||
| AK159369.1 | - | 12 | 1760 | |||
| AK170382.1 | - | 13 | 1971 | |||
| AK170827.1 | - | 13 | 2004 | |||
| AK170874.1 | - | 13 | 2014 | |||
| AK170881.1 | - | 13 | 1998 | |||
| AK171234.1 | - | 13 | 1999 | |||
| AK171411.1 | - | 13 | 2033 | |||
| AK171429.1 | - | 13 | 2011 | |||
| AK171479.1 | - | 13 | 2005 | |||
| AK171779.1 | - | 11 | 2879 | |||
| AK172562.1 | - | 13 | 2006 | |||
| AK172716.1 | - | 13 | 1958 | |||
| BC021477.1 | - | 13 | 1980 | 5343552 | ||
| BC034682.1 | - | 13 | 1924 | 3485807 | ||

Mical1
Genomic Map