siRNA Details
| Gene Symbol: Mc5r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013596.2 | NP_038624.2 | 2 | 441 | ||
| GenBank mRNA | BB650258.1 | - | 2 | 534 | ||
| BC100720.1 | - | 2 | 438 | 40045173 | ||
| BC100721.1 | - | 2 | 438 | 40045171 | ||
| BC100722.1 | - | 2 | 438 | 40045170 | ||
| BC100723.1 | - | 2 | 438 | 40045167 | ||
| U08354.1 | - | 2 | 447 | |||
More Information
Set Membership: |
|

Mc5r
Genomic Map