siRNA Details
| Gene Symbol: Mcf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159485.1 | NP_001152957.1 | 12 | 1636 | ||
| NM_001159486.1 | NP_001152958.1 | 12 | 1636 | |||
| NM_178076.3 | NP_835177.2 | 12 | 1539 | |||
| GenBank mRNA | AK122259.1 | - | 3 | 444 | ||
| AK147252.1 | - | 9 | 1743 | |||
| AK170186.1 | - | 12 | 1597 | |||
| AK170558.1 | - | 12 | 1570 | |||
| BC058622.1 | - | 12 | 1474 | 5704822 | ||
| DQ978380.1 | - | 11 | 1390 | |||
| S76838.1 | - | 13 | 1833 | |||

Mcf2l
Genomic Map