siRNA Details
| Gene Symbol: Cd46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010778.3 | NP_034908.1 | 2 | 307 | ||
| GenBank mRNA | AB001566.1 | - | 2 | 284 | ||
| AB010919.1 | - | 2 | 304 | |||
| AB022600.1 | - | 2 | 284 | |||
| AK006632.1 | - | 2 | 308 | |||
| BC150660.1 | - | 2 | 308 | 9087571 | ||

Cd46
Genomic Map