siRNA Details
| Gene Symbol: Anapc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008569.2 | NP_032595.2 | 33, 34 | 4496 | ||
| GenBank mRNA | AK034858.1 | - | 1, 2 | 27 | ||
| AK090134.1 | - | 33, 34 | 4478 | |||
| BC139002.1 | - | 33, 34 | 4477 | 8862024 | ||
| BC145492.1 | - | 33, 34 | 4477 | 9054055 | ||
| X80169.1 | - | 33, 34 | 4481 | |||

Anapc1
Genomic Map