siRNA Details
| Gene Symbol: Mdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162904.1 | NP_001156376.1 | 3, 4 | 674 | ||
| NM_010785.2 | NP_034915.2 | 3, 4 | 674 | |||
| NM_148922.3 | NP_683724.2 | 3, 4 | 674 | |||
| GenBank mRNA | AK004789.2 | - | 3, 4 | 629 | ||
| AK011769.1 | - | 3, 4 | 629 | |||
| BC052680.1 | - | 3, 4 | 624 | |||
| BC137608.1 | - | 3, 4 | 602 | 8860627 | ||
| BC144777.1 | - | 3, 4 | 602 | 9053325 | ||
| EU490427.1 | - | 3, 4 | 516 | |||
| M20823.1 | - | 3, 4 | 537 | |||
| M20824.1 | - | 3, 4 | 537 | |||

Mdm1
Genomic Map