siRNA Details
| Gene Symbol: Mdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF007110.1 | - | 11 | 1439 | ||
| AK017719.1 | - | 11 | 1413 | |||
| AK049576.1 | - | 11 | 1375 | |||
| AK049690.1 | - | 10 | 1340 | |||
| AK083349.1 | - | 12 | 1474 | |||
| AK139529.1 | - | 11 | 1349 | |||
| AK139535.1 | - | 11 | 1348 | |||
| BC003750.1 | - | 11 | 1418 | 3599906 | ||
| EU568360.1 | - | 12 | 1470 | |||

Mdm4
Genomic Map