siRNA Details
| Gene Symbol: Abcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008576.3 | NP_032602.1 | 28 | 4033 | ||
| GenBank mRNA | AF022908.1 | - | 28 | 3995 | ||
| AF213389.1 | - | 1 | 33 | |||
| AK029876.1 | - | 28 | 4034 | |||
| AK049174.1 | - | 12 | 1800 | |||
| AK140421.1 | - | 5 | 1576 | |||
| BC090617.1 | - | 28 | 4004 | 30943198 | ||

Abcc1
Genomic Map