siRNA Details
| Gene Symbol: Abcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008576.3 | NP_032602.1 | 23 | 3395 | ||
| GenBank mRNA | AF022908.1 | - | 23 | 3357 | ||
| AK029876.1 | - | 23 | 3396 | |||
| AK033750.1 | - | 1 | 178 | |||
| AK049174.1 | - | 7 | 1162 | |||
| AK140421.1 | - | 1 | 172 | |||
| BC090617.1 | - | 23 | 3366 | 30943198 | ||

Abcc1
Genomic Map