siRNA Details
| Gene Symbol: Abcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008576.3 | NP_032602.1 | 21 | 2854 | ||
| GenBank mRNA | AF022908.1 | - | 21 | 2816 | ||
| AK029876.1 | - | 21 | 2855 | |||
| AK049174.1 | - | 5 | 621 | |||
| BC090617.1 | - | 21 | 2825 | 30943198 | ||

Abcc1
Genomic Map