siRNA Details
| Gene Symbol: Mecom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021442.2 | NP_067417.1 | 2 | 208 | ||
| GenBank mRNA | AJ010015.1 | - | 2 | 208 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mecom| Gene Symbol: Mecom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021442.2 | NP_067417.1 | 2 | 208 | ||
| GenBank mRNA | AJ010015.1 | - | 2 | 208 | ||