siRNA Details
| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170537.1 | NP_001164008.1 | 8 | 1339 | ||
| NM_025282.3 | NP_079558.1 | 8 | 1333 | |||
| GenBank mRNA | AK009139.1 | - | 8 | 1173 | ||
| AK077603.1 | - | 7 | 1130 | |||
| AK132573.1 | - | 8 | 1168 | |||
| BC026841.1 | - | 7 | 1071 | 4481227 | ||
| BC037731.1 | - | 7 | 1061 | 4500786 | ||
| BC057650.1 | - | 7 | 1050 | 4500786 | ||
| L13171.1 | - | 6 | 900 | |||

Mef2c
Genomic Map