siRNA Details
Gene Symbol: Mef2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170537.1 | NP_001164008.1 | 8 | 1268 | ||
NM_025282.3 | NP_079558.1 | 8 | 1262 | |||
GenBank mRNA | AK009139.1 | - | 8 | 1102 | ||
AK132573.1 | - | 8 | 1097 | |||
BC026841.1 | - | 7 | 1000 | 4481227 | ||
BC037731.1 | - | 7 | 990 | 4500786 | ||
BC057650.1 | - | 7 | 979 | 4500786 | ||
L13171.1 | - | 6 | 829 |
