siRNA Details
| Gene Symbol: Fyco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110253.2 | NP_001103723.2 | 8 | 867 | ||
| NM_148925.3 | NP_683727.3 | 9 | 1114 | |||
| GenBank mRNA | AJ428065.1 | - | 9 | 1019 | ||
| AK081259.1 | - | 8 | 850 | |||
| BC053712.1 | - | 7 | 775 | |||

Fyco1
Genomic Map