siRNA Details
| Gene Symbol: Mep1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008585.2 | NP_032611.2 | 10 | 993 | ||
| GenBank mRNA | AK093914.1 | - | 10 | 952 | ||
| AK143906.1 | - | 10 | 951 | |||
| AK164734.1 | - | 10 | 948 | |||
| BC015258.1 | - | 10 | 993 | 4241252 | ||
| M74897.1 | - | 10 | 992 | |||

Mep1a
Genomic Map