siRNA Details
| Gene Symbol: Mep1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008586.2 | NP_032612.2 | 12 | 1709 | ||
| GenBank mRNA | AK098127.1 | - | 12 | 1709 | ||
| BC125627.1 | - | 12 | 1707 | 40130142 | ||
| BC145979.1 | - | 12 | 1708 | 40131282 | ||
| L15193.1 | - | 13 | 1709 | |||

Mep1b
Genomic Map