siRNA Details
| Gene Symbol: Mertk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008587.1 | NP_032613.1 | 10 | 1559 | ||
| GenBank mRNA | AK030158.1 | - | 2 | 263 | ||
| L11625.1 | - | 1 | 97 | |||
| U21301.1 | - | 10 | 1559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mertk| Gene Symbol: Mertk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008587.1 | NP_032613.1 | 10 | 1559 | ||
| GenBank mRNA | AK030158.1 | - | 2 | 263 | ||
| L11625.1 | - | 1 | 97 | |||
| U21301.1 | - | 10 | 1559 | |||