siRNA Details
| Gene Symbol: Mertk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008587.1 | NP_032613.1 | 3 | 546 | ||
| GenBank mRNA | U21301.1 | - | 3 | 546 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mertk| Gene Symbol: Mertk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008587.1 | NP_032613.1 | 3 | 546 | ||
| GenBank mRNA | U21301.1 | - | 3 | 546 | ||