siRNA Details
| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008591.2 | NP_032617.2 | 19 | 3929 | ||
| GenBank mRNA | BC117969.1 | - | 17 | 3573 | ||
| BC117970.1 | - | 17 | 3569 | |||
| BU514228.1 | - | 5 | 586 | |||
| Y00671.1 | - | 17 | 3574 | |||

Met
Genomic Map