siRNA Details
| Gene Symbol: Mid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290504.1 | NP_001277433.1 | 802 | |||
| NM_001290505.1 | NP_001277434.1 | 1026 | ||||
| NM_001290506.1 | NP_001277435.1 | 912 | ||||
| NM_001290512.1 | NP_001277441.1 | 246 | ||||
| NM_010797.3 | NP_034927.2 | 909 | ||||
| GenBank mRNA | AF026565.1 | - | 802 | |||
| AK135734.1 | - | 1025 | ||||
| AK160120.1 | - | 248 | ||||
| AY540038.1 | - | 214 | ||||
| AY540040.1 | - | 214 | ||||
| AY540041.1 | - | 100 | ||||
| AY540042.1 | - | 214 | ||||
| BC053704.1 | - | 879 | 30058736 | |||
| Y14848.1 | - | 863 | ||||

Mid1
Genomic Map