siRNA Details
| Gene Symbol: Mitf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113198.1 | NP_001106669.1 | 9 | 1203 | ||
| NM_001178049.1 | NP_001171520.1 | 9 | 1090 | |||
| NM_008601.3 | NP_032627.1 | 8 | 894 | |||
| GenBank mRNA | AB221657.1 | - | 8 | 756 | ||
| AK052895.1 | - | 9 | 1062 | |||
| AK154189.1 | - | 9 | 1204 | |||
| AK155350.1 | - | 9 | 1204 | |||
| BC108976.2 | - | 8 | 858 | 40047440 | ||
| BC108977.1 | - | 8 | 823 | 40047441 | ||
| L22958.1 | - | 1 | 46 | |||
| Z23066.1 | - | 8 | 885 | |||

Mitf
Genomic Map