siRNA Details
| Gene Symbol: Mki67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081117.2 | NP_001074586.2 | 12 | 2969 | ||
| GenBank mRNA | AK052772.1 | - | 12 | 2889 | ||
| AK082099.1 | - | 12 | 2888 | |||
| AK143677.1 | - | 12 | 3008 | |||
| X82786.1 | - | 12 | 3077 | |||

Mki67
Genomic Map