siRNA Details
| Gene Symbol: Aff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080798.1 | NP_001074267.1 | 3 | 489 | ||
| NM_133919.3 | NP_598680.3 | 3 | 500 | |||
| GenBank mRNA | AF013131.1 | - | 2 | 315 | ||
| AF074266.1 | - | 3 | 333 | |||
| AK082878.1 | - | 4 | 557 | |||
| AK142554.1 | - | 3 | 500 | |||
| AK143742.1 | - | 3 | 489 | |||
| AY749168.1 | - | 3 | 333 | |||
| BC131665.1 | - | 3 | 333 | 40110795 | ||

Aff1
Genomic Map