siRNA Details
| Gene Symbol: Mmp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008605.3 | NP_032631.3 | 8 | 1167 | ||
| GenBank mRNA | AK089309.1 | - | 8 | 1136 | ||
| AK089452.1 | - | 8 | 1136 | |||
| AK089509.1 | - | 9 | 1010 | |||
| AK089523.1 | - | 8 | 1136 | |||
| AK090051.1 | - | 8 | 1138 | |||
| AK150596.1 | - | 8 | 1136 | |||
| AK151021.1 | - | 8 | 1136 | |||
| AK151273.1 | - | 8 | 1136 | |||
| AK151933.1 | - | 8 | 1136 | |||
| AK155139.1 | - | 8 | 1135 | |||
| AK155743.1 | - | 8 | 1136 | |||
| AK155747.1 | - | 8 | 1136 | |||
| AK155748.1 | - | 8 | 1136 | |||
| AK155841.1 | - | 8 | 1194 | |||
| AK155865.1 | - | 8 | 1135 | |||
| AK170360.1 | - | 8 | 1136 | |||
| AK170364.1 | - | 8 | 1136 | |||
| AK170500.1 | - | 9 | 1010 | |||
| AK170667.1 | - | 7 | 2184 | |||
| AK170798.1 | - | 8 | 1135 | |||
| AK170804.1 | - | 8 | 1135 | |||
| AK170976.1 | - | 8 | 1136 | |||
| AK171006.1 | - | 8 | 1136 | |||
| AK171032.1 | - | 8 | 1135 | |||
| AK171036.1 | - | 8 | 1136 | |||
| AK171176.1 | - | 8 | 1136 | |||
| AK171428.1 | - | 8 | 1136 | |||
| AK171435.1 | - | 7 | 2183 | |||
| AK171447.1 | - | 8 | 1136 | |||
| AK171448.1 | - | 8 | 1136 | |||
| AK171449.1 | - | 8 | 1136 | |||
| AK171450.1 | - | 8 | 1136 | |||
| AK171466.1 | - | 8 | 1136 | |||
| AK171468.1 | - | 8 | 1136 | |||
| AK171470.1 | - | 8 | 1136 | |||
| AK171472.1 | - | 8 | 1136 | |||
| AK171485.1 | - | 8 | 1136 | |||
| AK171518.1 | - | 8 | 1136 | |||
| AK171542.1 | - | 8 | 1136 | |||
| AK171570.1 | - | 8 | 1136 | |||
| BC019135.1 | - | 8 | 1130 | 5037674 | ||
| CT010377.1 | - | 8 | 1083 | |||
| M82831.1 | - | 8 | 1083 | |||

Mmp12
Genomic Map