siRNA Details
Gene Symbol: Mmp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008609.3 | NP_032635.1 | 2 | 776 | ||
GenBank mRNA | BC047278.1 | - | 2 | 776 | 6390113 | |
BC057952.1 | - | 2 | 346 | 5342023 | ||
D86332.1 | - | 2 | 272 |
