siRNA Details
| Gene Symbol: Mmp24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010808.3 | NP_034938.3 | 5 | 988 | ||
| GenBank mRNA | AB021226.1 | - | 5 | 933 | ||
| AJ010262.1 | - | 5 | 934 | |||
| CD352405.1 | - | 2 | 185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp24| Gene Symbol: Mmp24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010808.3 | NP_034938.3 | 5 | 988 | ||
| GenBank mRNA | AB021226.1 | - | 5 | 933 | ||
| AJ010262.1 | - | 5 | 934 | |||
| CD352405.1 | - | 2 | 185 | |||