siRNA Details
| Gene Symbol: Clec4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163161.1 | NP_001156633.1 | 6 | 831 | ||
| NM_010819.4 | NP_034949.3 | 6 | 834 | |||
| GenBank mRNA | AF061272.1 | - | 6 | 766 | ||
| AK089500.1 | - | 6 | 825 | |||
| AK150926.1 | - | 6 | 737 | |||
| AK155919.1 | - | 6 | 753 | |||
| AK170692.1 | - | 6 | 695 | |||
| AK171409.1 | - | 6 | 772 | |||
| BC099471.1 | - | 6 | 744 | 30845162 | ||

Clec4d
Genomic Map