siRNA Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018736.2 | NP_061206.1 | 6 | 573 | ||
| GenBank mRNA | AK041248.1 | - | 6 | 572 | ||
| AK141214.1 | - | 6 | 585 | |||
| AK146234.1 | - | 6 | 606 | |||
| AK146370.1 | - | 6 | 606 | |||
| AK168180.1 | - | 6 | 606 | |||
| AK168208.1 | - | 6 | 607 | |||
| BC065144.1 | - | 6 | 573 | 30627207 | ||
| U58987.1 | - | 6 | 608 | |||
| U60318.1 | - | 6 | 608 | |||

Mre11a
Genomic Map