siRNA Details
| Gene Symbol: Msh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008628.2 | NP_032654.1 | 2 | 334 | ||
| GenBank mRNA | AK051960.1 | - | 2 | 331 | ||
| AK078775.1 | - | 2 | 319 | |||
| AK157842.1 | - | 2 | 347 | |||
| AK166179.1 | - | 2 | 320 | |||
| BC047117.1 | - | 2 | 342 | 4951173 | ||
| BC050897.1 | - | 2 | 317 | 6401835 | ||
| U21011.1 | - | 2 | 351 | |||
| X81143.1 | - | 2 | 334 | |||

Msh2
Genomic Map