siRNA Details
| Gene Symbol: Msh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010830.2 | NP_034960.1 | 3 | 640 | ||
| GenBank mRNA | AK088259.1 | - | 3 | 629 | ||
| BC035274.1 | - | 1 | 38 | |||
| BC051160.1 | - | 3 | 585 | 4953238 | ||
| BC051634.1 | - | 3 | 595 | 6518577 | ||
| CN458717.1 | - | 3 | 652 | |||
| U42190.1 | - | 3 | 616 | |||

Msh6
Genomic Map