siRNA Details
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010832.5 | NP_034962.2 | 8 | 1156 | ||
GenBank mRNA | AF117066.2 | - | 8 | 1158 | ||
AK077794.1 | - | 2 | 416 | |||
AK089368.1 | - | 8 | 1239 | |||
AK146208.1 | - | 8 | 1092 | |||
AK148898.1 | - | 8 | 1134 | |||
BC010226.1 | - | 8 | 1117 | 3501126 |
