siRNA Details
| Gene Symbol: Mtm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164190.1 | NP_001157662.1 | 8 | 740 | ||
| NM_001164191.1 | NP_001157663.1 | 8 | 631 | |||
| NM_001164192.1 | NP_001157664.1 | 8 | 631 | |||
| NM_019926.3 | NP_064310.2 | 8 | 620 | |||
| GenBank mRNA | AF073996.1 | - | 8 | 621 | ||
| AK016445.1 | - | 8 | 741 | |||
| AK031794.1 | - | 8 | 602 | |||
| AK166452.1 | - | 8 | 599 | |||
| AK171475.1 | - | 8 | 609 | |||
| BC051022.1 | - | 8 | 589 | |||
| BC066847.1 | - | 8 | 578 | |||
| BC090984.1 | - | 8 | 598 | 30246206 | ||

Mtm1
Genomic Map