siRNA Details
| Gene Symbol: Laptm4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008640.2 | NP_032666.2 | 4 | 1277 | ||
| GenBank mRNA | AK013963.1 | - | 3 | 295 | ||
| AK075699.1 | - | 4 | 512 | |||
| AK084515.1 | - | 4 | 920 | |||
| AK088958.1 | - | 4 | 537 | |||
| AK165638.1 | - | 4 | 546 | |||
| AK184706.1 | - | 4 | 321 | |||
| BC086667.1 | - | 4 | 457 | 30633914 | ||
| BC089021.1 | - | 4 | 490 | 6734038 | ||
| U34259.1 | - | 5 | 1144 | |||

Laptm4a
Genomic Map