siRNA Details
| Gene Symbol: Prol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008644.2 | NP_032670.2 | 3 | 708 | ||
| GenBank mRNA | U37531.1 | - | 4 | 591 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prol1| Gene Symbol: Prol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008644.2 | NP_032670.2 | 3 | 708 | ||
| GenBank mRNA | U37531.1 | - | 4 | 591 | ||