siRNA Details
| Gene Symbol: Mut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008650.3 | NP_032676.2 | 5, 6 | 1206 | ||
| GenBank mRNA | AK146309.1 | - | 5, 6 | 1166 | ||
| BC019175.1 | - | 5, 6 | 1188 | 5007694 | ||
| X51941.1 | - | 4, 5 | 1117 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mut| Gene Symbol: Mut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008650.3 | NP_032676.2 | 5, 6 | 1206 | ||
| GenBank mRNA | AK146309.1 | - | 5, 6 | 1166 | ||
| BC019175.1 | - | 5, 6 | 1188 | 5007694 | ||
| X51941.1 | - | 4, 5 | 1117 | |||