siRNA Details
| Gene Symbol: Mvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF137598.1 | - | 2, 3 | 235 | ||
| AK009261.1 | - | 4, 5 | 345 | |||
| AK181570.1 | - | 1, 2 | 89 | |||
| AK187523.1 | - | 1, 2 | 89 | |||
| BC005606.1 | - | 3, 4 | 288 | 3498140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mvk| Gene Symbol: Mvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF137598.1 | - | 2, 3 | 235 | ||
| AK009261.1 | - | 4, 5 | 345 | |||
| AK181570.1 | - | 1, 2 | 89 | |||
| AK187523.1 | - | 1, 2 | 89 | |||
| BC005606.1 | - | 3, 4 | 288 | 3498140 | ||