siRNA Details
| Gene Symbol: Mx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010846.1 | NP_034976.1 | 1474 | |||
| GenBank mRNA | BC011113.1 | - | 1459 | 4011821 | ||
| CT010406.1 | - | 1261 | ||||
| M12279.1 | - | 1474 | ||||
| M21038.1 | - | 1546 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mx1| Gene Symbol: Mx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010846.1 | NP_034976.1 | 1474 | |||
| GenBank mRNA | BC011113.1 | - | 1459 | 4011821 | ||
| CT010406.1 | - | 1261 | ||||
| M12279.1 | - | 1474 | ||||
| M21038.1 | - | 1546 | ||||