siRNA Details
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010846.1 | NP_034976.1 | 10 | 2085 | ||
GenBank mRNA | AK076430.1 | - | 14 | 1837 | ||
AK150023.1 | - | 9 | 1565 | |||
AK150236.1 | - | 9 | 1565 | |||
BC011113.1 | - | 9 | 2070 | 4011821 | ||
CT010406.1 | - | 1872 | ||||
EU638327.1 | - | 5 | 890 | |||
EU638328.1 | - | 8 | 1383 | |||
M12279.1 | - | 10 | 2085 | |||
M21038.1 | - | 11 | 2157 | |||
M21039.1 | - | 11 | 1661 |
