siRNA Details
| Gene Symbol: Mxi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008542.2 | NP_001008542.2 | 5 | 895 | ||
| NM_001008543.2 | NP_001008543.1 | 5 | 582 | |||
| NM_010847.3 | NP_034977.1 | 5 | 735 | |||
| GenBank mRNA | AK140465.1 | - | 5 | 685 | ||
| AK144219.1 | - | 5 | 777 | |||
| AK154538.1 | - | 5 | 885 | |||
| BC064453.1 | - | 5 | 880 | 6810025 | ||
| D31824.1 | - | 5 | 735 | |||
| L38821.1 | - | 5 | 588 | |||
| L38822.1 | - | 5 | 607 | |||
| U24673.1 | - | 5 | 568 | |||
| U24674.1 | - | 5 | 517 | |||

Mxi1
Genomic Map