siRNA Details
| Gene Symbol: Mybl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008652.2 | NP_032678.1 | 1, 2 | 224 | ||
| GenBank mRNA | AK028497.1 | - | 1, 2 | 224 | ||
| BC050842.1 | - | 1, 2 | 170 | 6518298 | ||
| X70472.1 | - | 1, 2 | 172 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mybl2| Gene Symbol: Mybl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008652.2 | NP_032678.1 | 1, 2 | 224 | ||
| GenBank mRNA | AK028497.1 | - | 1, 2 | 224 | ||
| BC050842.1 | - | 1, 2 | 170 | 6518298 | ||
| X70472.1 | - | 1, 2 | 172 | |||