siRNA Details
| Gene Symbol: Myh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161775.1 | NP_001155247.1 | 38 | 5463 | ||
| NM_013607.2 | NP_038635.2 | 38 | 5463 | |||
| GenBank mRNA | AK173045.1 | - | 38 | 5427 | ||
| BC026142.1 | - | 38 | 5448 | 3989495 | ||
| D85923.1 | - | 38 | 5418 | |||
| D85924.1 | - | 38 | 5418 | |||

Myh11
Genomic Map