siRNA Details
| Gene Symbol: Myo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103171.1 | NP_001096641.1 | 61 | 10143 | ||
| NM_010862.2 | NP_034992.2 | 62 | 10197 | |||
| NM_182698.2 | NP_874357.2 | 60 | 6381 | |||
| GenBank mRNA | AF144095.1 | - | 62 | 10197 | ||
| AK030528.1 | - | 12 | 1941 | |||
| AY331132.1 | - | 61 | 10143 | |||
| AY331133.1 | - | 60 | 6381 | |||

Myo15
Genomic Map