siRNA Details
| Gene Symbol: Myo15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103171.1 | NP_001096641.1 | 3 | 3865 | ||
| NM_010862.2 | NP_034992.2 | 3 | 3865 | |||
| NM_182698.2 | NP_874357.2 | 2 | 103 | |||
| GenBank mRNA | AF053130.1 | - | 2 | 77 | ||
| AF144095.1 | - | 3 | 3865 | |||
| AY331132.1 | - | 3 | 3865 | |||
| AY331133.1 | - | 2 | 103 | |||

Myo15
Genomic Map