siRNA Details
| Gene Symbol: Myo7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032394.3 | NP_115770.2 | 26, 27 | 3520 | ||
| GenBank mRNA | AF242411.1 | - | 26, 27 | 3557 | ||
| AK220152.1 | - | 1, 2 | 101 | |||
| BC138338.1 | - | 26, 27 | 3520 | 8861358 | ||
| BC138341.1 | - | 26, 27 | 3520 | 8861361 | ||

Myo7b
Genomic Map