siRNA Details
| Gene Symbol: Myo7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032394.3 | NP_115770.2 | 37 | 5156 | ||
| GenBank mRNA | AF242411.1 | - | 37 | 5193 | ||
| AK220152.1 | - | 12 | 1737 | |||
| BC026416.1 | - | 10 | 1500 | |||
| BC138338.1 | - | 37 | 5156 | 8861358 | ||
| BC138341.1 | - | 37 | 5156 | 8861361 | ||

Myo7b
Genomic Map