siRNA Details
| Gene Symbol: Myoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010865.3 | NP_034995.3 | 1 | 504 | ||
| GenBank mRNA | AB013592.1 | - | 1 | 488 | ||
| AF039869.1 | - | 1 | 485 | |||
| AK144395.1 | - | 1 | 500 | |||
| BC125329.1 | - | 1 | 449 | 40129844 | ||
| BC137606.1 | - | 1 | 449 | 8860625 | ||

Myoc
Genomic Map