siRNA Details
| Gene Symbol: Myt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171615.1 | NP_001165086.1 | 7 | 869 | ||
| NM_001171616.1 | NP_001165087.1 | 7 | 1093 | |||
| NM_001171680.1 | NP_001165151.1 | 1 | 1059 | |||
| NM_008665.4 | NP_032691.2 | 7 | 1093 | |||
| GenBank mRNA | AB082378.1 | - | 7 | 1080 | ||
| AB093267.2 | - | 7 | 800 | |||
| AF004294.1 | - | 1 | 1060 | |||
| BC063252.1 | - | 7 | 869 | 6406740 | ||

Myt1
Genomic Map